missing translation for 'onlineSavingsMsg'
Learn More

Palmatine chloride, MedChemExpress

Product Code. 16459895
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459895 250 mg 250mg
16432499 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16459895 Supplier Medchem Express Supplier No. HYN0110
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Palmatine chloride is an orally active and irreversible indoleamine 2,3-dioxygenase 1 (IDO-1) inhibitor with IC50s of 3 μM and 157μM against HEK 293-hIDO-1 and rhIDO-1, respectively.

Palmatine chloride can also inhibit West Nile virus (WNV) NS2B-NS3 protease in an uncompetitive manner with an IC50 of 96 μM. Palmatine chloride shows anti-cancer, anti-oxidation, anti-inflammatory, neuroprotection, antibacterial, anti-viral activities.

Specifications

Chemical Name or Material Palmatine chloride
CAS 10605-02-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H22ClNO4
Quantity 250 mg
Solubility Information DMSO : ≥ 32 mg/mL (82.50 mM) ∣H2O : 5 mg/mL (12.89 mM; Need ultrasonic)
SMILES COC1=C(OC)C2=C[N+]3=C(C4=CC(OC)=C(OC)C=C4CC3)C=C2C=C1.[Cl-]
Molecular Weight (g/mol) 387.86
Formula Weight 387.86
Percent Purity 96.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.