missing translation for 'onlineSavingsMsg'
Learn More

Palmitelaidic Acid, TRC

Product Code. 30412957
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412957 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412957 Supplier Toronto Research Chemicals Supplier No. TRCP144005250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Palmitelaidic Acid
CAS 10030-73-6
Molecular Formula C16 H30 O2
Quantity 250 mg
Synonym 9-Hexadecenoic acid','(E)- (8CI), (9E)-9-Hexadecenoic acid, (E)-9-Hexadecenoic acid, (E)-Hexadec-9-enoic acid, 9-trans-Hexadecenoic acid, Palmitelaidic acid, trans-9-Hexadecenoic acid, trans-Palmitoleic acid, 9-Hexadecenoic acid','(9E)-, (E)-9-Hexadecenoic Acid
InChI Formula InChI=1S/C16H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h7-8H,2-6,9-15H2,1H3,(H,17,18)/b8-7+
SMILES CCCCCC\C=C\CCCCCCCC(=O)O
IUPAC Name (E)-hexadec-9-enoic acid
Molecular Weight (g/mol) 254.41
Formula Weight 254.2246
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.