missing translation for 'onlineSavingsMsg'
Learn More

Palmitoyl chloride, 98%

Product Code. 10668021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10668021 100 mL Glass bottle 100mL
10731161 5 mL Glass bottle 5mL
10013690 500 mL Glass bottle 500mL
10217463 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10668021 Supplier Thermo Scientific Acros Supplier No. 304291000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-67-4 | C16H31ClO | 274.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-67-4
MDL Number MFCD00000742
InChI Key ARBOVOVUTSQWSS-UHFFFAOYSA-N
Synonym palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride
PubChem CID 8206
IUPAC Name hexadecanoyl chloride
SMILES CCCCCCCCCCCCCCCC(=O)Cl

Specifications

Acidity 1% max. (as palmitic acid)
Melting Point 9.0°C to 14.0°C
Density 0.9000g/mL
Boiling Point 199.0°C (2.0 mmHg)
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C16H31ClO
Linear Formula CH3(CH2)14COCl
Refractive Index 1.4510 to 1.4530
Quantity 100 mL
Beilstein 02, 374
Specific Gravity 0.9
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ether,decomposes in alcohol
Molecular Weight (g/mol) 274.88
Formula Weight 274.88
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Palmitoyl chloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Palmitoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : Hexadecanoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.