missing translation for 'onlineSavingsMsg'
Learn More

Palmityl palmitate, 96%

Product Code. 15495548
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15495548 5 g 5g
15485548 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15495548

Brand: Thermo Scientific Alfa Aesar H59283.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 540-10-3 | C32H64O2 | 480.86 g/mol

It is used as emollients, masking agents and skin conditioning and a glosser and thickener for creams. It improves emulsion texture and stability and gives structure to cosmetic sticks. Used in skin and hair applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as emollients, masking agents and skin conditioning and a glosser and thickener for creams. It improves emulsion texture and stability and gives structure to cosmetic sticks. Used in skin and hair applications.

Solubility
Soluble in hot acetone.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-10-3
Molecular Formula C32H64O2
Molecular Weight (g/mol) 480.86
MDL Number MFCD00053739
InChI Key PXDJXZJSCPSGGI-UHFFFAOYSA-N
Synonym cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester
PubChem CID 10889
ChEBI CHEBI:75584
IUPAC Name hexadecyl hexadecanoate
SMILES CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC

Specifications

Melting Point 55°C to 56°C
Linear Formula CH3(CH2)14CO2(CH2)15CH
Quantity 5 g
Solubility Information Soluble in hot acetone.
Formula Weight 480.85
Percent Purity 96%
Chemical Name or Material Palmityl palmitate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.