missing translation for 'onlineSavingsMsg'
Learn More

Palmityl Palmitate, TRC

Product Code. 30136898
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136898 5 g 5g
30134686 1 g 1g
30005726 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136898 Supplier Toronto Research Chemicals Supplier No. TRCP1637505G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-10-3
Molecular Formula C32 H64 O2
Molecular Weight (g/mol) 480.85
Synonym Cetyl Palmitate
IUPAC Name hexadecyl hexadecanoate
SMILES CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C32H64O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-34-32(33)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-31H2,1-2H3
Formula Weight 480.4906
Chemical Name or Material Cetyl palmitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.