missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ Palonosetron hydrochloride

Product Code. 18785386
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
missing translation for 'unitSize':
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
18785386 50 mg 50mg
18777396 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18785386 missing translation for 'mfr' Tocris™ missing translation for 'supplierNo' 7038/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Highly potent and high affinity 5-HT3 antagonist

Palonosetron hydrochloride is a highly potent and high affinity 5-HT3 antagonist (IC50 = 0.24 nM for 5-HT3A; pKi = 10.4). Displays a slow dissociation rate. Inhibits 5-HT-induced enhancement of cellular response to substance P (Cat. No. 1156) in vitro. Inhibits cisplatin (Cat. No. 2251)-induced enhancement of substance P response in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3 AS)-2-[(3 S)-1-azabicyclo[2.2.2 ]oct-3-yl]-2,3,3 A,4,5,6-hexahydro-1 H-benz[de]isoquinolin-1-one hydrochloride
CAS 135729-62-3
Quantity 50 mg
Target 5-HT3 Receptor Antagonists
Molecular Formula C19H24N2O.HCl
Formula Weight 2778
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.