missing translation for 'onlineSavingsMsg'
Learn More

Pamidronic Acid, TRC

Product Code. 30136914
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136914 100 mg 100mg
30134797 250 mg 250mg
30009881 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136914 Supplier Toronto Research Chemicals Supplier No. TRCP172490100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40391-99-9
Molecular Formula C3 H11 N O7 P2
Molecular Weight (g/mol) 235.07
Synonym Phosphonic acid, P,P'-(3-amino-1-hydroxypropylidene)bis-, Phosphonic acid, (3-amino-1-hydroxypropylidene)bis- (9CI), P,P'-(3-Amino-1-hydroxypropylidene)bis[phosphonic acid], (3-Amino-1-hydroxypropylidene)-1,1-bisphosphonate, (3-Amino-1-hydroxypropylidene)bisphosphonic acid, (α-Hydroxy-γ-aminopropylidene)diphosphonic acid, 1-Hydroxy-3-aminopropane-1,1-diphosphonic acid, 3-Amino-1-hydroxypropane-1,1-diphosphonic acid, 3-Amino-1-hydroxypropylidene-1,1-bisphosphonic acid, 3-Amino-1-hydroxypropylidenediphosphonic acid, ADP, AHPrBP, Amidronic acid, BPH 5, Pamidronic acid, Panolin, Propane-1-hydroxy-3-amino-1,1-diphosphonic acid
IUPAC Name (3-amino-1-hydroxy-1-phosphonopropyl)phosphonic acid
SMILES NCCC(O)(P(=O)(O)O)P(=O)(O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C3H11NO7P2/c4-2-1-3(5,12(6,7)8)13(9,10)11/h5H,1-2,4H2,(H2,6,7,8)(H2,9,10,11)
Formula Weight 235.0011
Chemical Name or Material Pamidronic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.