missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Panorama Safety Goggles

Product Code. 11961651 Shop All Bürkle Products
Change view
Click to view available options
Material:
Polycarbonate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
11961651 Polycarbonate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11961651 Supplier Buerkle™ Supplier No. 25021001
  • ONLINE EXCLUSIVE PRICE
    18.90 €
    16.20 € / Pack of 1
    Save 2.70 €
    14% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A very comfortable pair of over-goggles with an unrestricted side view.

  • Overspecs for prescription wearers
  • Very good all-round vision
  • Length-adjustable temples for a pressure-free fit
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DIN EN 166 - 168 and 170, DIN 4646/47
Lens Coating Uncoated
Color Translucent
Lens Tint None
Product Type Safety Glasses
For Use With (Application) Eye Protection
Frame Color Clear
No. per Pack 1 Ea.
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Vented Side Arms
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.