Learn More
Motic™ Panthera Cloud Binocular Digital Microscope
Product Code. 17609830
Change view
Click to view available options
Objective Types:
Plan Achromat Objectives UC (4X, 10X, 40X(S), 100X(S-oil))
Unit Size:
Each
Product Code. Objective Types Unit Size
17609830 Plan Achromat Objectives UC (4X, 10X, 40X(S), 100X(S-oil)) Each
1 options
Product Code. 17609830 Supplier Motic™ Supplier No. 1100104601181
  • ONLINE EXCLUSIVE PRICE
    3690.00 €
    3268.37 € / Pack of 1
    Save 421.63 €
    11% Off
View all promotions
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Panthera Cloud has smartly been created to make your workflow easier with strong Ultra Contrast optics.
  • 25° inclined binocular Siedentopf head
  • Rotates 360°
  • SmartCam 5 MP camera
  • Strong optical performance with Ultra Contrast (UC) optics
  • Diopter Adjustment: ±4
Specifications
Eyepiece WF 10X/22 mm
Head Style Binocular
Lighting Type Kohler Illumination
Magnification Power 10X
Objective Types Plan Achromat Objectives UC (4X, 10X, 40X(S), 100X(S-oil))