missing translation for 'onlineSavingsMsg'
Learn More

Pantoprazole, TRC

Product Code. 30135976
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135976 1 g 1g
30135908 250 mg 250mg
17901215 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135976 Supplier Toronto Research Chemicals Supplier No. TRCP1900031G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102625-70-7
Molecular Formula C16 H15 F2 N3 O4 S
Molecular Weight (g/mol) 383.37
Synonym (±)-Pantoprazole, 1H-Benzimidazole, 6-(difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]sulfinyl]-, 1H-Benzimidazole, 5-(difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]sulfinyl]- (9CI), 6-(Difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole, (±)-Pantoprazole, 2-[[(3,4-Dimethoxypyridin-2-yl)methyl]sulfinyl]-5-difluoromethoxy-1H-benzimidazole, 5-(Difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole, 5-(Difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridyl)methyl]sulfinyl]-1H-benzimidazole, Altopan, BY 1023, Pantocid, Pantopan, Pantoprazole, Pantozol, Protium, Protium (pharmaceutical), SKF 96022, Zovanta
IUPAC Name 5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methylsulfinyl]-1H-benzimidazole
SMILES COc1ccnc(CS(=O)c2nc3cc(OC(F)F)ccc3[nH]2)c1OC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H15F2N3O4S/c1-23-13-5-6-19-12(14(13)24-2)8-26(22)16-20-10-4-3-9(25-15(17)18)7-11(10)21-16/h3-7,15H,8H2,1-2H3,(H,20,21)
Formula Weight 383.0751
Percent Purity >95%
Chemical Name or Material Pantoprazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.