missing translation for 'onlineSavingsMsg'
Learn More

(+)-Pantothenic Acid Calcium Salt, Extra Pure, SLR, Fisher Chemical

Product Code. 10467070 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10467070 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10467070 Supplier Fisher Chemical Supplier No. P/0295/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 190°C
CAS 137-08-6
Color White
pH 6.8 to 8.0
Physical Form Solid
Quantity 100 g
Molecular Formula C18H32CaN2O10
MDL Number MFCD00002766
Synonym calcium pantothenate, d-pantothenic acid hemicalcium salt, d-pantothenic acid, calcium salt, pantothenic acid, calcium salt, d, r-n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-beta-alanine calcium salt, beta-alanine, n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-, calcium salt, r
InChI Key FAPWYRCQGJNNSJ-DXHDTSSINA-L
SMILES [Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O
IUPAC Name calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoic acid
Molecular Weight (g/mol) 476.54
PubChem CID 131847364
Formula Weight 476.52g/mol
Packaging Plastic powder jar
Density Vapor Density: 16.4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.