missing translation for 'onlineSavingsMsg'
Learn More

Pantothenic Acid Hemicalcium Salt, TRC

Product Code. 30135065
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135065 100 g 100g
30137560 25 g 25g
17937354 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135065 Supplier Toronto Research Chemicals Supplier No. TRCP190300100G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-08-6
Molecular Formula 2 C9 H16 N O5 . Ca
Molecular Weight (g/mol) 476.53
Synonym D-Pantothenic Acid Calcium, β-Alanine, N-[(2R)-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl]-, calcium salt (2:1), Calcium bis[3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoate], Calcium, bis(pantothenato)- (7CI), Pantothenic acid, calcium salt (2:1), D- (8CI), β-Alanine, N-(2,4-dihydroxy-3,3-dimethyl-1-oxobutyl)-, calcium salt (2:1), (R)-, (+)-Pantothenic acid calcium salt, Calcium D-(+)-N-(α,γ-dihydroxy-β,β-dimethylbutyryl)-β-alaninate, Calcium D-(+)-pantothenate, Calcium D-pantothenate, Calcium pantothenate, Calpan, Calpanate, Dextro calcium pantothenate, N-(2,4-Dihydroxy-3,3-dimethylbutyryl)-β-alanine calcium, NSC 36292, Pancal, Panthoject, Pantholin, Panthothenic acid hemicalcium salt, Pantothenate calcium, Pantothenic acid calcium salt, Pantothenic acid hemicalcium salt, Vitamin B5 calcium salt
IUPAC Name calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoate
SMILES [Ca+2].CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)[O-].CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)[O-]

Specifications

InChI Formula InChI=1S/2C9H17NO5.Ca/c2*1-9(2,5-11)7(14)8(15)10-4-3-6(12)13;/h2*7,11,14H,3-5H2,1-2H3,(H,10,15)(H,12,13);/q;;+2/p-2/t2*7-;/m00./s1
Quantity 100 g
Formula Weight 476.1683
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Calcium Pantothenate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.