missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Papain

Product Code. 16489206
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489206 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16489206 Supplier MedChemExpress Supplier No. HYP1645
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Papain is a cysteine protease of the peptidase C1 family, which is used in food, pharmaceutical, textile, and cosmetic industries.

Specifications

CAS 9001-73-4
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) COVID-19-immunoregulation
Product Type Natural Products
Quantity 100 mg
Shipping Condition Room temperature
Solubility H2O : 50 mg/mL (Need ultrasonic) ∣DMSO : 5 mg/mL (Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.