missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Papain, Carica Papaya Latex

Product Code. 15404369
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15404369 25 mg 25mg
15414369 100 mg 100mg
15494359 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15404369 Supplier Thermo Scientific Chemicals Supplier No. J61875.MB
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A cysteine protease; breaks down intercellular matrix of cartilage | CAS: 9001-73-423 kd g/mol

Papain, from papaya latex, has been used to digest carotid artery smooth muscle cells. Papain is used in dissecting solutions. It is used to produce Fab fragments of antibodies. It is used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9001-73-4
Synonym EC 3.4.22.2

Specifications

For Use With (Application) A cysteine protease; breaks down intercellular matrix of cartilage
Molecular Weight (g/mol) 23 kd
Content And Storage 2°C to 8°C
Form Lyophilized powder,23 kDa
Quantity 25 mg
Product Type Protease
compliance-icons compliance-icons
Product Identifier
  • Papain, Carica Papaya Latex
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Papain

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.