missing translation for 'onlineSavingsMsg'
Learn More

Papaveraldine, TRC

Product Code. 30008823
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008823 100 mg 100mg
30003839 1 g 1g
30011477 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008823 Supplier Toronto Research Chemicals Supplier No. TRCP190400100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 522-57-6
Molecular Formula C20 H19 N O5
Molecular Weight (g/mol) 353.3
Synonym NSC 94266, Papaveraldin, 6,7-Dimethoxy-1-veratroylisoquinoline;
IUPAC Name (6,7-dimethoxyisoquinolin-1-yl)-(3,4-dimethoxyphenyl)methanone
SMILES COc1ccc(cc1OC)C(=O)c2nccc3cc(OC)c(OC)cc23

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H19NO5/c1-23-15-6-5-13(10-16(15)24-2)20(22)19-14-11-18(26-4)17(25-3)9-12(14)7-8-21-19/h5-11H,1-4H3
Formula Weight 353.3686
Chemical Name or Material Papaveraldine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.