Learn More
Brady™ Paper Labels for M6/M7 Printers
Paper Labels with Permanent Rubber Adhesive provide good contrast and smear resistance for thermal transfer and barcode printing applications.
Brand:  Brady™ 173383
 View more versions of this product
Product Code. 17818854
  • ONLINE EXCLUSIVE PRICE
    26.50 €
    25.49 € / Pack of 100
    Save 1.01 €
    3% Off
In Stock
Add to basket
Description
Description
  • White matte paper label stock features permanent rubber adhesive
  • Low cost is ideal for all general purpose labeling projects
  • Service temperature of -40°C to 50°C (-40°F to 122°F)
  • Made with B-424 permanent adhesive
Specifications
Specifications
Not Autoclavable
Permanent Rubber
White
Die-cut
Single-side Printable
Label
Barcode Labeling, Electrical Labeling, General Identification, Laboratory Labeling
No
Rectangle
-40°C to +50°C
0.089 mm
2 in.
25.4 mm
M7-20-424
ASTM D 1000
None
Matte
Paper
Thermal Transfer
Non-sterile
M710, BMP71
100 Labels
-40°F to +122°F
0.0035 in.
2 x 1 in.
1 in.
50.8 mm