missing translation for 'onlineSavingsMsg'
Learn More

Amcor™ Parafilm™ M Laboratory Film

Product Code. 11772644
Change view
Click to view available options
Length (Metric):
15.2 m
38 m
75 m
76 m
Width (Metric):
5 cm
10 cm
51 cm
Color:
Natural
Purple
Unit Size:
Each
Pack of 24
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Color Unit Size
11772644 38 m 10 cm Natural Each
11747487 15.2 m 51 cm Natural Each
10018130 76 m 10 cm Natural Each
11762644 76 m 5 cm Natural Each
11693119 76 m 5 cm Natural Pack of 24
16330422 75 m 5 cm Purple Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11772644 Supplier Amcor™ Supplier No. PM996
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Efficient and cost-effective covering for test tubes, beakers, vials, petri dishes, flasks, and various other instruments and objects with irregular surfaces.

  • Protect the integrity of research applications with a tight moisture-barrier that holds a b seal and doesn't release from glassware, despite off-gassing
  • Reduce the risk of tampered data with a covering that is odorless, colorless and semi-transparent
  • Prevents moisture and volume loss due to material composition and tight seal
  • Flexible and self-sealing material allows it to cling around irregular shapes and surfaces
  • Stretches more than 200% of original length
  • Resistant up to 48 hours against many polar substances such as saline solutions, inorganic acids and alkaline solutions

Permeability

  • Oxygen: (ASTM 1927-98) 150 cc/m2 d at 23°C and 50% RH
  • Carbon Dioxide: (Modulated IR Method) 1200 cc/m2 d at 23°C and 0% RH
  • Water Vapor: (ASTM F1249-01)
    • Flat: 1 g/m2 d at 38°C and 90% RH
    • Creased: 1 g/m2 d at 38°C and 90% RH

Effects of Common Reagents

  • Potassium Permanganate
    • 5%: No apparent effect except permanent dark brown coloration in 18 hours
    • 0.1%: Same as 5% except slightly less color
  • Iodine solution (0.1N)
    • No effect except staining brown in 18 hours
  • Ethyl Alcohol (95%)
    • No apparent effect except some face whitening in 24 hours
  • No apparent effect in 24 hours:
    • Hydrochloric Acid conc. (12N) dil. (5N)
    • Sulphuric Acid conc. (36N) dil. (5N)
    • Nitric Acid conc. (16N) dil. (5N)
    • Sodium Hydroxide conc. (22%)
    • Ammonium Hydroxide conc. (28% NH3)
    • Salt (NaCl) solution (20%)
    • Isopropyl Alcohol (99%)
  • Not recommended for use with chlorinated, non-polar aliphatic and aromatic solvents
  • Film becomes soft and sticky at about 130° to 150°F (54° to 66°C)

Order Info

13-374-10, 13-374-12, and 13-374-16 rolls are packaged in cardboard dispensing cartons.

Specifications

Applications Efficient and cost-effective covering, over wrap to further secure stoppers, lids, and caps, and shelf or tray liner to prevent slipping of bottles or instruments.
Length (English) 125 ft.
Color Natural
Description Amcor PM996
Product Type Film
Width (English) 4 in.
Width (Metric) 10 cm
Length (Metric) 38 m
Material Thermoplastic
Thickness 5 mil
For Use With (Equipment) Parafilm Wrap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.