missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Paraformaldehyde, 4% in PBS + Mg + EGTA

Product Code. 15454859
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454859 500 mL 500mL
15444859 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454859 Supplier Thermo Scientific Chemicals Supplier No. J62478.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as a common fixative reagent for histology. It is also used to prepare pure formaldehyde. | CAS: 30525-89-4 | CH2O | 30.026 g/mol

Paraformaldehyde, 4% in PBS + Mg + EGTA is used as a common fixative reagent for histology. It is also used to prepare pure formaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30525-89-4
Molecular Formula CH2O
Molecular Weight (g/mol) 30.026
MDL Number MFCD00133991
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Physical Form Liquid
Packaging Plastic bottle
Quantity 500 mL
Solubility Information Immiscible with water.
Chemical Name or Material Paraformaldehyde
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Paraformaldehyde, 4% in PBS + Mg + EGTA
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Paraformaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.