missing translation for 'onlineSavingsMsg'
Learn More

Paraformaldehyde, Extra Pure, SLR, Fisher Chemical

Product Code. 10131580 Shop All Fisher Chemical Products
Change view
Click to view available options
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Unit Size
10131580 Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10131580 Supplier Fisher Chemical Supplier No. P/0840/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

500g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

CAS 30525-89-4
Melting Point 120°C/248°F
Color White
pH 3.5 to 5.0
Packaging Plastic powder jar
Molecular Formula CH2O
MDL Number 133991
Quantity 500 g
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILES C=O
IUPAC Name formaldehyde
Molecular Weight (g/mol) 30.026
PubChem CID 712
ChEBI CHEBI:16842
Vapor Pressure 1.2mmHg 25
Physical Form Solid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Paraformaldehyde
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2
  • Acute toxicity Category 4
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.