missing translation for 'onlineSavingsMsg'
Learn More

Paraformaldehyde, MP Biomedicals™

Product Code. 11496572 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496572 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11496572 Supplier MP Biomedicals Supplier No. 02150146.5
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as a disinfectant and fumigant. Paraformaldehyde, MP Biomedicals is the smallest polyoxymethylene, a polymerization product of formaldehyde. It is also used in the preparation of formalin fixatives for tissues or cells when the samples are to be used

Paraformaldehyde is used as a fixative in electron microscopy and in the preparation of formalin fixatives for tissues or cells when the samples are to be used in florescence studies. It is also used in manufacturing synthetic resins. It has been used in the past for disinfecting sickrooms, clothing, linen and sickroom utensils, and as a fumigant.

Specifications

CAS 30525-89-4
Melting Point 120–170°C (Lit.)
Density 0.88g/mL at 25°C (Lit.)
Boiling Point Does not boil, gives off formaldehyde gas when heated
Molecular Formula CH2O
Quantity 500 g
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
Solubility Information Soluble in water (hot water - ∼37% w/v; slowly soluble in cold water). Insoluble in alcohol or ether.
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILES C=O
IUPAC Name formaldehyde
Molecular Weight (g/mol) 30.026
PubChem CID 712
ChEBI CHEBI:16842
Formula Weight (30.03)n
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.