missing translation for 'onlineSavingsMsg'
Learn More

Paraformaldehyde Solution, 16%, Methanol Free, MP Biomedicals™

Product Code. 11481745 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2 x 10 mL
10 x 10 mL
Unit Size:
100mL
20mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481745 10 x 10 mL 100mL
15799150 2 x 10 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481745 Supplier MP Biomedicals Supplier No. 0219998380
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MP Biomedicals™ Paraformaldehyde Solution, 16%, Methanol Free is a clear, aqueous solution that is ready to use as a fixative for electron microscopy.

Recommended for use in the preparation of formalin fixatives for tissues or cells when the samples are to be used in florescence studies.

Chemical Identifiers

CAS 30525-89-4 , 7732-18-5

Specifications

CAS Max % 0.16, 0.84
Molecular Formula (CH2O)n
Quantity 10 x 10 mL
Synonym Polyoxmethylene, POM
Percent Purity 0.16
Grade Methanol free
Concentration or Composition (by Analyte or Components) 16% aqueous solution
Physical Form Liquid
Chemical Name or Material Paraformaldehyde Solution
compliance-icons compliance-icons
Product Identifier
  • Paraformaldehyde, 16% aq soln, methanol free
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Carcinogenicity Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302+H312+H332-Harmful if swallowed, in contact with skin or if inhaled
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: paraformaldehyd
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.