missing translation for 'onlineSavingsMsg'
Learn More

Paraformaldehyde, TRC

Product Code. 30012467
Change view
Click to view available options
Quantity:
50 g
100 g
150 g
Unit Size:
100g
150g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012467 150 g 150g
30006728 50 g 50g
30005087 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012467 Supplier Toronto Research Chemicals Supplier No. TRCP193265150G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30525-89-4
Molecular Formula (C H2 O)n
Molecular Weight (g/mol) 30.03
Synonym Paraformaldehyde (9CI, ACI), Aldacide, Flo-Mor, Granuform, Granuform 91, Granuform 96, Granuform M, Paraform, PFA, Polyformaldehyde, TransFix, WANPFA 92
IUPAC Name Paraformaldehyde
SMILES [*]CO[*]

Specifications

Quantity 150 g
InChI Formula InChI=1S/CH2O/c1-2/h1H2
Formula Weight 30.0106
Chemical Name or Material Paraformaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.