missing translation for 'onlineSavingsMsg'
Learn More

Paraldehyde, TRC

Product Code. 30135883
Change view
Click to view available options
Quantity:
25 mL
100 mL
250 mL
Unit Size:
100mL
250mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135883 100 mL 100mL
30133021 25 mL 25mL
30004525 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135883 Supplier Toronto Research Chemicals Supplier No. TRCP191820100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-63-7
Molecular Formula C6 H12 O3
Molecular Weight (g/mol) 132.16
Synonym Paraldehyde, 2,4,6-Trimethyl-1,3,5-trioxane, cyclic trimer of acetaldehyde
IUPAC Name 2,4,6-trimethyl-1,3,5-trioxane
SMILES CC1OC(C)OC(C)O1

Specifications

Quantity 100 mL
InChI Formula InChI=1S/C6H12O3/c1-4-7-5(2)9-6(3)8-4/h4-6H,1-3H3
Formula Weight 132.0786
Chemical Name or Material Paraldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.