missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Parameter Immunoassay Control Set 916 for Human B2M

Product Code. 18733519
Change view
Click to view available options
Content And Storage:
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Unit Size
18733519 Store the unopened product at 2°C to 8°C. Do not use past expiration date. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18733519 Supplier R&D Systems™ Supplier No. QC224
  • ONLINE EXCLUSIVE PRICE
    223.04 €
    198.45 € / Pack of 1
    Save 24.59 €
    11% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Immunoassay control set for Human beta 2-Microglobulin assay

beta 2-Microglobulin (B2M) forms the light chain of Class I MHC molecules. It has been measured in a variety of body fluids, including serum/plasma, saliva, CSF, and urine. B2M freely passes through the glomerular membrane, but it is nearly completely reabsorbed and degraded in proximal tubule cells. Clinically, serum B2M is elevated in rheumatoid arthritis, systemic lupus erythematosus, viral infections, and conditions associated with decreased glomerular filtration.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Type Control Set for ELISA Kit
Gene ID (Entrez) 567 (Human); 12010 (Mouse); 24223 (Rat)
Intended Use For quantitative measurement of beta 2 microglobulin levels
Product Line R&D Systems
Product Name Parameter Immunoassay Control Set
Source Human
Target Gene beta 2-Microglobulin

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.