Learn More
Labconco™ Paramount™ Ductless Enclosure Accessory, Telescoping Base Stand
For use with Paramount™ Ductless Enclosure. Labconco™ Paramount™ Ductless Enclosure Accessory, Telescoping Base Stand with casters may be adjusted to eight height positions prior to installation.
Brand:  Labconco™ 3746710
Product Code. 15119655
  • 1520.00 € / Each
Description
  • Durable 4.4cm (1.75 in.) tubular, powder-coated steel frame
  • Casters provide mobility for short distances
  • Telescoping legs are adjustable to eight height positions in 2.54cm (1 in.) increments from 69.8 to 87.6cm (27.5 to 34.5 in.) (must be adjusted prior to installation)
  • Includes four 12.7cm (5 in.) diameter, toe-locking, non-marking polyurethane casters with bearings for quiet operation
  • Models 91.4cm (3 ft.) wide and larger have rear crossbar supports
  • Support Paramount Ductless Enclosures and other select Labconco enclosures
  • Work surface is required (sold separately)
  • Models with four fixed leveling feet also avilable
  • 16-gauge steel accessory shelves are available separately to hook on lower rear cross member for storage and supplies