missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pararosaniline chloride, 96%, pure

Product Code. 11946931 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11946931 100 g Glass Bottle 100g
10746003 5 g Glass Bottle 5g
10507182 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11946931 Supplier Thermo Scientific Chemicals Supplier No. 416801000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 569-61-9 | C19H17N3·HCl | 323.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ideal for analysis by ICP-MS and high-resolution ICP-MS.
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569-61-9
Molecular Formula C19H17N3·HCl
Molecular Weight (g/mol) 323.83
InChI Key JUQPZRLQQYSMEQ-UHFFFAOYSA-N
Synonym basic red 9, pararosaniline hydrochloride, parafuchsin, pararosaniline chloride, parafuchsine, p-fuchsin, basic fuchsin, ci basic red 9, c.i. basic red 9, parafuksin
PubChem CID 11292
ChEBI CHEBI:87663
IUPAC Name 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl

Specifications

Melting Point 268.0°C to 270.0°C
Color Green
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 97.5% min. (HPLC)
UN Number 1873
Solubility Information Solubility in water: 10g/L (25°C). Other solubilities: 65g/L ethanol
Formula Weight 323.83
Percent Purity 95%
Grade Pure
Infrared Spectrum Authentic
Packaging Glass Bottle
Chemical Name or Material Pararosaniline Chloride
Show More Show Less
compliance-icons
Product Identifier
  • Pararosaniline chloride
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.