missing translation for 'onlineSavingsMsg'
Learn More

CELLPATH PARASHIELD

Product Code. 30226328
Change view
Click to view available options
Quantity:
5 x 250 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30226328 5 x 250 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30226328 Supplier CELLPATH Supplier No. XWA020000A
  • ONLINE EXCLUSIVE PRICE
    368.02 €
    170.51 € / Pack of 5
    Save 197.51 €
    53% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ParaShield is a precisely formulated wax repellent manufactured by CellPath. It can not only be used to stop paraffin from adhering to laboratory counter tops, but also to clean or act as a barrier on metal, wood and plastic surfaces in areas where paraffin is likely to be used.

  • To apply ParaShield, simply spray onto a cloth and then apply a thin layer to the relevant surface.
  • This will not only dissolve any paraffin already present but also provide an immediate protective barrier to potential sticking.
  • It is not recommended for use on items that come in direct contact with ribbons, such as microtome blades, blade holders or base moulds, due to ParaShield's paraffin dissolving nature.
  • Key Benefits
  • Stops paraffin adhering to laboratory surfaces and equipment
  • Xylene free and non toxic repellent with a pleasant smell
  • Suitable for use on multiple surfaces, including metal, wood and plastic

Specifications

Physical Form Liquid
Color Clear Colorless
Quantity 5 x 250 mL
Grade Laboratory
compliance-icons
Product Identifier
  • Parashield
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Lytol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.