Learn More
Emma Safety Footwear Paris Safety Shoes
Product Code. 15998401
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15998401 43 1 pair
15958401 39 1 pair
15968401 40 1 pair
15978401 41 1 pair
15988401 42 1 pair
15908411 44 1 pair
15918411 45 1 pair
15928411 46 1 pair
15938411 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15998401 Supplier Emma Safety Footwear Supplier No. MM40464743
  • ONLINE EXCLUSIVE PRICE
    145.37 €
    100.12 € / 1 pairs
    Save 45.25 €
    31% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Lightweight woven, polyurethane-coated textile with Nano Carbon Composite protective toe. Performs excellently at workplaces where safety and long-lasting comfort are top priorities. The extremely comfortable PU insole guarantees long-lasting support and cushioning of the foot.
  • S1P-Safety shoe with sole plate for dry work
  • Woven Textile
  • Polyurethane coated
  • Hydro-Tec™ Antibacterial
  • Metal-free
  • Chrome-free lining
  • ESD certified
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black, Gray
Design Paris
Liner Material Hydro-Tec™ Antibacterial
Material Textile
Size 43
Toe Material Composite