missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paromomycin sulfate, MedChemExpress
Description
Paromomycin (Aminosidine) sulfate, a neomycin (HY-B0470) derivative, is a broad spectrum aminoglycoside antibiotic with amebicidal and bactericidal effects. Paromomycin sulfate prematures termination of translation of mRNA and inhibits protein synthesis by specifically binds to the RNA oligonucleotide at the A site of bacterial 30S ribosomes. Paromomycin sulfate can be used for the research of bacterial and parasitic infections.
Specifications
Specifications
| Chemical Name or Material | Paromomycin sulfate |
| CAS | 1263-89-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H47N5O18S |
| Quantity | 1 g |
| Synonym | Aminosidine sulfate |
| Solubility Information | H2O : 100 mg/mL (140.11 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : 1 mg/mL (1.40 mM; ultrasonic and warming and heat to 80°C) ∣Ethanol : 1 mg/mL (1.40 mM; Need ultrasonic) |
| SMILES | N[C@@H](C[C@H]1N)[C@@]([C@@H]([C@H]1O)O[C@@](O[C@H](CO)[C@H]2O[C@@]([C@@H]([C@@H](O)[C@@H]3O)N)([H])O[C@H]3CN)([H])[C@@H]2O)([H])O[C@H]([C@@H]([C@@H](O)[C@@H]4O)N)O[C@@H]4CO.O=S(O)(O)=O |
| Molecular Weight (g/mol) | 713.71 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?