missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Paroxetine hydrochloride hemihydrate, 98%

Product Code. 15873855 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15873855 5 g 5g
15863855 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15873855 Supplier Thermo Scientific Chemicals Supplier No. 462630050
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
This item is not returnable. View return policy

CAS: 110429-35-1 | C19H20FNO3 | 329.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110429-35-1
Molecular Formula C19H20FNO3
Molecular Weight (g/mol) 329.37
InChI Key AHOUBRCZNHFOSL-YOEHRIQHSA-N
IUPAC Name (3S,4R)-3-[(2H-1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine
SMILES FC1=CC=C(C=C1)[C@@H]1CCNC[C@H]1COC1=CC=C2OCOC2=C1

Specifications

Color Clear Colorless
Assay Percent Range 97.5 % min. (HPLC)
Quantity 5 g
Solubility Information Solubility aq. soln.: very slightly soluble. Other solubilities: soluble in methanol, dmf, ethanol
Formula Weight 374.83
Chemical Name or Material Paroxetine hydrochloride hemihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.