missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Particle-Certified Glass Containers

Product Code. 15798927
Change view
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
2500 mL
4000 mL
Product Type:
Boston Round Bottle
Jug
Packer Bottle
Sample Jar
Color:
Amber
Clear
No. per Case:
12/Cs.
24/Cs.
4/Cs.
Unit Size:
Each
Pack of 12
Pack of 24
Pack of 4
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Product Type Color No. per Case Unit Size
15798927 250 mL Packer Bottle Amber 12/Cs. Pack of 12
15311757 60 mL Boston Round Bottle Amber 24/Cs. Pack of 24
15930371 125 mL Boston Round Bottle Amber 12/Cs. Pack of 12
16474390 125 mL Boston Round Bottle Clear 12/Cs. Pack of 12
15535205 250 mL Boston Round Bottle Clear 24/Cs. Pack of 24
16875781 500 mL Sample Jar Clear 12/Cs. Each
15935322 500 mL Boston Round Bottle Amber 12/Cs. Pack of 12
16484390 1000 mL Sample Jar Clear 12/Cs. Pack of 12
15926195 2500 mL Packer Bottle Amber 4/Cs. Pack of 4
15916195 4000 mL Jug Amber 4/Cs. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 15798927 Supplier Thermo Scientific™ Supplier No. 12108A/LP
  • ONLINE EXCLUSIVE PRICE
    484.00 €
    257.00 € / Pack of 12
    Save 227.00 €
    46% Off
Estimated Shipment: 24-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.

Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.

Key Features:

  • Class 100/10 glass
  • Certificate of Analysis is included with each shipment
  • Available in clear or amber
  • Assembled with low-shedding polypropylene septa with chemically inert PTFE faced liners that do not contain adhesives
  • Individual bottles double bagged to maintain integrity until point of use
  • 4 L jugs suitable for hazardous shipping in conjunction with combination packaging

Recommended for:

  • Low particle applications
TRUSTED_SUSTAINABILITY

Specifications

Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Color Amber
Description Amber Glass Wide Mouth Packer Bottle with Closure, 250 mL
Diameter (English) 4.75 in.
Material Glass
No. per Case 12/Cs.
Closure Included Yes
Graduated No
Mouth Wide
Height (Metric) 63.5 mm
Height (English) 2.5 in.
Closure Liner PTFE
Closure Material Polypropylene, Low-shedding
Closure Size 45-400 mm
Diameter (Metric) 120.65 mm
Autoclavable Autoclavable
Capacity (English) 8 oz.
Shape Round
Product Type Packer Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.