missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences pASK-IBA E. coli vector

Product Code. 12816486 Shop All IBA Lifesciences Products
Change view
Click to view available options
Vector:
pASK-IBA2C
pASK-IBA3C
pASK-IBA4C
pASK-IBA5C
pASK-IBA6C
pASK-IBA7C
Unit Size:
5µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Vector Unit Size
12816486 pASK-IBA3C 5µg
12806486 pASK-IBA2C 5µg
12826486 pASK-IBA4C 5µg
12836486 pASK-IBA5C 5µg
12846486 pASK-IBA6C 5µg
12856486 pASK-IBA7C 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 12816486 Supplier IBA Lifesciences Supplier No. 21322000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

pASK-IBA E. coli expression vectors encoding a Strep-tag™II and chloramphenicol resistance

pASK-IBA vectors are classic cloning vectors devised for periplasmic or cytoplasmic E. coli expression of recombinant proteins with the Strep-tag™II fused to the C- or N-terminus. They carry the chloramphenicol resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, a multiple cloning site with recognition sites for several common restriction enzymes (e.g. EcoRI, SmaI, BamHI, SalI, PstI) and the inducible tetracycline promoter/operator for the regulated expression of proteins. Some carry the ompA signal for periplasmic secretion of the recombinant protein.The vector can be combined with any E. coli strain since the tet-promoter works independently of the genetic background of E. coli. Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity. During secretion into the periplasmic space the ompA signal sequence will be cleaved off.

Specifications

Format Suspension
Promoter Tet
Storage Buffer TE buffer
Antibiotic Resistance Bacterial Chloramphenicol
Expression System E. coli
Protein Tag Position (to your gene) C-terminal
Replication Origin F1 and ColE1 origin
Size Range approx. 3 kb
Protein Tag Strep-tag II
Cloning Method Direct Cloning
Concentration 250 ng/μL
Vector pASK-IBA3C
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.