Learn More
Microspec™ Pasteur Pipette
Product Code. 16625452
Change view
Click to view available options
Volume (Metric) Dispense:
1 mL
3 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Dispense Sterility unitSize
16625452 1 mL Non-sterile Pack of 500
16655452 1 mL Sterile Pack of 500
16635452 3 mL Non-sterile Pack of 500
16645452 3 mL Sterile Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16625452 Supplier Microspec™ Supplier No. AC1
In Stock
Add to basket
Edge
Add to basket
Device used to transfer small quantities of liquids
Bulk Pack of 500. Available as sterile or non-sterile.
Specifications
Autoclavable Not Autoclavable
Calibrated No
Sterility Non-sterile
Graduated Yes
Quantity 500
Volume (Metric) Dispense 1 mL
Color Clear
Disposable Single-use