missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ Jurkat ICOS Signaling Assay
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The PathHunter ICOS Checkpoint Signaling cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting signaling checkpoints. This cell line enables assessment of ligand (e.g. ICOS-L) based activation of ICOS receptors via detection of SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged ICOS target protein and EA-tagged SH2 domain protein. Upon activation of the receptor by its ligand or an agonist antibody, recruitment of SH2 domain protein to the phosphorylated receptor tail leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Jurkat |
| Cell Line | Jurkat |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Anti-ICOS antibody |
| Quantity | 2 Vials |
| Target Entry | ICOS |
| Accession Number | NM_012092.3 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction