missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ U2OS NF-KB Pathway Reporter Cell Line

Product Code. 17334429
Encompass
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
This item is not returnable. View return policy

Product Code. 17334429

Brand: DiscoverX 931157C3

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ U2OS NF-kB Pathway Reporter Cell Line measures NF-kB pathway activation through detection of induced expression levels of a tagged reporter protein. This cell line ships as two vials of cryopreserved cells.

The PathHunter™ U2OS NF-kB Pathway Reporter Cell Line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting the NFkB pathway. This cell line enables assessment of NFkB pathway stimulation based on changes in the expression of a NFkB response element-driven reporter protein. This product uses EFC technology. The included cell line expresses PL-tagged reporter protein under the control of a NFkB pathway-inducible transcriptional response element. Pathway activation induces expression of the PL-tagged protein, allowing detection of EFC signal upon addition of EA-containing detection reagent. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Transcriptional Reporter
Cell Line U2OS
For Use With (Application) Cell Lines
Primary Agonist CD40L
Quantity 2 Vials
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.