missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS p53 Nuclear Translocation Cell Line
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The PathHunter™ U2OS p53 Nuclear Translocation Cell Line is a stable, engineered cell line for use in studying drug candidates affecting the p53 DNA damage pathway. This cell line enables assessment of ligand (e.g. camptothecin) based pathway activation via detection of resulting p53 nuclear translocation. This product uses EFC technology. The included cell line overexpresses PL-tagged p53 and nuclear localized EA alongside untagged Keap1. Upon pathway activation, induced nuclear translocation of p53 leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | p53 Nuclear Translocation |
| Cell Line | U2OS |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Camptothecin |
| Quantity | 2 Vials |
| Target Entry | p53 |
| Accession Number | NM_000546.2 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction