missing translation for 'onlineSavingsMsg'
Learn More

Patulin, 99%

Product Code. 10572821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Packaging:
Glass bottle
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10572821 10 mg Glass bottle 10mg
10013773 5 mg Glass bottle 5mg
10194900 25 mg Glass bottle 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10572821 Supplier Thermo Scientific Acros Supplier No. 228870100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 149-29-1 | C7H6O4 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-29-1
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00005858
InChI Key ZRWPUFFVAOMMNM-UHFFFAOYSA-N
Synonym patulin, clavacin, expansin, clavatin, claviform, expansine, patuline, clairformin, claviformin, gigantin
PubChem CID 4696
ChEBI CHEBI:74926
IUPAC Name 4-hydroxy-4,6-dihydrofuro[3,2-c]pyran-2-one
SMILES C1C=C2C(=CC(=O)O2)C(O1)O

Specifications

Melting Point 108.0°C to 111.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 mg
Merck Index 15, 7158
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents,soluble in ethyl and amyl acetate,insoluble in petroleum ether
Formula Weight 154.12
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Patulin, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Patulin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.