Learn More
Emma Safety Footwear Paul Safety Shoes
Product Code. 15998831
Change view
Click to view available options
Size:
37
38
39
40
41
44
45
46
47
48
Unit Size:
1 pair
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15998831 45 1 pair
15958831 37 1 pair
15968831 38 1 pair
15795233 39 1 pair
15978831 40 1 pair
15705243 41 1 pair
15988831 44 1 pair
15908841 46 1 pair
15918841 47 1 pair
15928841 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15998831 Supplier Emma Safety Footwear Supplier No. MM70084645
  • ONLINE EXCLUSIVE PRICE
    143.14 €
    83.95 € / 1 pairs
    Save 59.19 €
    41% Off
Estimated Shipment: 18-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
An ideal choice when optimal foot protection is required. A strong, solid steel plate in the sole and resilient leather uppers defend the feet from unexpected hazards. Perfectly balanced, anatomical arch support will keep feet feeling fit and fresh.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black
Design Paul
Liner Material Hydro-Tec™ Sanitized Silver
Material Leather
Size 45
Toe Material Steel