missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emma Safety Footwear Paul Safety Shoes
An ideal choice when optimal foot protection is required. A strong, solid steel plate in the sole and resilient leather uppers defend the feet from unexpected hazards. Perfectly balanced, anatomical arch support will keep feet feeling fit and fresh.
120.00 € - 120.84 €
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Closure Type | Laces |
Color | Black |
Design | Paul |
Liner Material | Hydro-Tec™ Sanitized Silver |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15958831
|
Emma Safety Footwear
MM70084637 |
37 |
120.84 €
1 pair |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
15968831
|
Emma Safety Footwear
MM70084638 |
38 |
120.84 €
1 pair |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
15795233
|
Emma Safety Footwear
MM70084639 |
39 |
120.00 €
1 pair |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
15978831
|
Emma Safety Footwear
MM70084640 |
40 |
120.84 €
1 pair |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
15705243
|
Emma Safety Footwear
MM70084641 |
41 |
120.00 €
1 pair |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
15988831
|
Emma Safety Footwear
MM70084644 |
44 |
120.84 €
1 pair |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
15998831
|
Emma Safety Footwear
MM70084645 |
45 |
120.84 €
1 pair |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
15908841
|
Emma Safety Footwear
MM70084646 |
46 |
120.84 €
1 pair |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
15918841
|
Emma Safety Footwear
MM70084647 |
47 |
120.84 €
1 pair |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
15928841
|
Emma Safety Footwear
MM70084648 |
48 |
120.84 €
1 pair |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Duo Heat Resistant Outsole
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
Steel |
Laces | |
Paul | |
Leather |