missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Paxilline

Product Code. 30001306
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001306 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001306 Supplier R&D Systems™ Supplier No. 2006/10
  • ONLINE EXCLUSIVE PRICE
    452.21 €
    404.25 € / 10mg
    Save 47.96 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. SERCA ATPase blocker. Also potent BKCa channel blocker

Potent blocker of high-conductance Ca2+-activated K+ (BKCa, KCa1.1) channels. Binds to the α-subunit of BKCa (Ki = 1.9 nM for block of currents in α-subunit-expressing oocytes) and enhances binding of charybdotoxin to BKCa channels in vascular smooth muscle. Also inhibits sarco/endoplasmic reticulum Ca2+-ATPase (IC50 = 5 - 50 μM).
TRUSTED_SUSTAINABILITY

Specifications

CAS 57186-25-1
Quantity 10 mg
Product Line Tocris
Molecular Formula C27H33NO4
Formula Weight 435.56
Purity >98%
Molecular Weight (g/mol) 435.564
Chemical Name or Material Paxilline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.