missing translation for 'onlineSavingsMsg'
Learn More

Paxilline, 97+%, Thermo Scientific Chemicals

Product Code. 15433949
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433949 5 mg 5mg
15443949 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15433949 Supplier Thermo Scientific Alfa Aesar Supplier No. J61341.#0
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
This item is not returnable. View return policy

Potent blocker of high-conductance calcium-activated potassium (BKCa) channels

Potent blocker of high-conductance calcium-activated potassium (BKCa) channels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57186-25-1
Molecular Formula C27H33NO4
Molecular Weight (g/mol) 435.564
MDL Number MFCD00083464
InChI Key ACNHBCIZLNNLRS-UBGQALKQSA-N
Synonym paxilline, unii-3t9u9z96l7, 2h-1-benzopyrano 5',6':6,7 indeno 1,2-b indol-3 4bh-one, 5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-, 2-alpha,4b-beta,6a-alpha,12b-beta,12c-alpha,14a-beta, 2h-pyrano 2,3:5',6' benz 1',2':6,7 indeno 1,2-b indol-3 4bh-one, 5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-, 2r,4bs,6as,12bs,12cr,14as, spectrum5_001975, paxilline, powder hplc, paxilline solution, 100 mug/ml in acetonitrile, analytical standard, 2r,4bs,6as,12bs,12cr,14as-5,6,6a,7,12,12b,12c,13,14,14a-decahydro-4b-hydroxy-2-1-hydroxy-1-methylethyl-12b,12c-dimethyl-2h-pyrano 2,3:5',6' benz 1',2':6,7 indeno 1,2-b indol-3 4bh-one, ncgc00025342-07_c27h33no4_ 2r,4bs,6as,12bs,12cr,14as-4b-hydroxy-2-2-hydroxy-2-propanyl-12b,12c-dimethyl-5,6,6a,7,12,12b,12c,13,14,14a-decahydro-2h-chromeno 5',6':6,7 indeno 1,2-b indol-3 4bh-one
PubChem CID 105008
ChEBI CHEBI:34907
SMILES CC12CCC3C(=CC(=O)C(O3)C(C)(C)O)C1(CCC4C2(C5=C(C4)C6=CC=CC=C6N5)C)O

Specifications

Color White to Yellow
Quantity 5 mg
UN Number UN2811
Solubility Information Soluble in DMSO,acetone or chloroform.
Formula Weight 435.56
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material Paxilline
compliance-icons compliance-icons
Product Identifier
  • Paxilline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Paxilline from Penicillium paxilli

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.