missing translation for 'onlineSavingsMsg'
Learn More

Pazufloxacin Mesylate, TRC

Product Code. 30414835
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414835 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414835 Supplier Toronto Research Chemicals Supplier No. TRCP2110001G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pazufloxacin Mesylate
CAS 163680-77-1
Molecular Formula C16 H15 F N2 O4 . C H4 O3 S
Quantity 1 g
Synonym 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid','10-(1-aminocyclopropyl)-9-fluoro-2,3-dihydro-3-methyl-7-oxo-','(3S)-','methanesulfonate (1:1), 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid','10-(1-aminocyclopropyl)-9-fluoro-2,3-dihydro-3-methyl-7-oxo-','(3S)-','monomethanesulfonate (9CI), 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid','10-(1-aminocyclopropyl)-9-fluoro-2,3-dihydro-3-methyl-7-oxo-','(S)-','monomethanesulfonate, Pasil, Pazucross, Pazufloxacin mesilate, Pazufloxacin mesylate, Pazufloxacin methanesulfonate, T 3762
InChI Formula InChI=1S/C16H15FN2O4.CH4O3S/c1-7-6-23-14-11(16(18)2-3-16)10(17)4-8-12(14)19(7)5-9(13(8)20)15(21)22;1-5(2,3)4/h4-5,7H,2-3,6,18H2,1H3,(H,21,22);1H3,(H,2,3,4)/t7-;/m0./s1
SMILES C[C@H]1COc2c3N1C=C(C(=O)O)C(=O)c3cc(F)c2C4(N)CC4.CS(=O)(=O)O
Molecular Weight (g/mol) 414.41
Formula Weight 414.0897
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.