Learn More
Thermo Scientific PC Thickwall Tube
Product Code. 15540084 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Centrifuges
Unit Size:
Pack of 10
Product Code. For Use With (Equipment) unitSize
15540084 Centrifuges Pack of 10
1 options
Product Code. 15540084 Supplier Thermo Scientific Supplier No. 75000610
Add to basket
Add to basket
PC Thickwall Tube for micro-ultracentrifuge.
  • PC Thickwall tube, 25 mL x 10, for micro-ultracentrifuge
TRUSTED_SUSTAINABILITY
Specifications
Material Polycarbonate
For Use With (Equipment) Centrifuges
Disposable Reusable
Capacity 10 x 25 mL
Description PC Thickwall Tube, 25 mL x 10, for micro-ultracentrifuge
Product Type Thickwall Tube