missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ pcDNA™3.4 TOPO™ TA Cloning Kit

Product Code. 13473269
Change view
Click to view available options
Quantity:
20 μg Kit
Unit Size:
10 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13473269 20 μg Kit 10 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13473269 Supplier Gibco™ Supplier No. A14697

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in suspension-adapted cells for transient protein expression. Gibco™ pcDNA™3.4 TOPO™ TA Cloning Kit is a constitutive mammalian expression vector designed to deliver exceptionally high levels of transgene expression.

  • Used as a Geneticin™ selectable expression plasmid for engineering stable cell lines
  • Full length human cytomegalovirus (CMV) immediate-early promoter/enhancer for high level gene expression in a wide range of mammalian cells
  • WPRE (woodchuck posttranscriptional regulatory element) downstream of the cloning site to enhance transcript expression
  • TOPO™ cloning site for rapid and efficient (>85%) cloning of Taq-amplified PCR products
  • Neomycin resistance gene for selection of stable cell lines with Geneticin™
  • pUC origin for high copy replication and maintenance of the plasmid in E. coli
TRUSTED_SUSTAINABILITY

Specifications

Antibiotic Resistance Bacterial Ampicillin (AmpR)
Cloning Method TOPO™-TA
Constitutive or Inducible System Constitutive
Content And Storage Store pcDNA™3.4-TOPO™ vector and all cloning reagents at -20°C.
Store One Shot™ TOP10 Chemically Competent E. coli cells at -80°C.
Delivery Type Transfection
For Use With (Application) Protein Production, Constitutive Expression
Plasmid High Copy Plasmid
Product Line TOPO
Product Type TA Cloning Kit
Promoter CMV
Protein Tag Untagged
Quantity 20 μg Kit
Replication Origin pUC Origin
Selection Agent (Eukaryotic) Geneticin™ (G-418)
Selection Marker Eukaryotic NeoR
Selection Marker Promoter SV40 Promoter
Vector pcDNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.