missing translation for 'onlineSavingsMsg'
Learn More

PCR Markers, Thermo Scientific Chemicals

Product Code. 15649008 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 μL
Unit Size:
250µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15649008 250 μL 250µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15649008 Supplier Thermo Scientific Supplier No. J76710.&M
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

6X DNA Loading Buffer (OXG) with orange G and xylene cyanol FF

 

  • Mixture of 8 DNA bands ranging in size from 50 to 2000 bp
  • Recommended amount for loading is 5μL per gel lane
  • 6X DNA Loading Buffer (OXG) with orange G and xylene cyanol FF is included for convenience to prepare PCR samples for agarose gel electrophoresis
  • Tracking dyes do not interfere with UV illumination of ethidium bromide stained gels
  • Fragments: 2000 bp; 1500 bp; 1000 bp; 750 bp; 500 bp; 300 bp; 150 bp; 50 bp
TRUSTED_SUSTAINABILITY

Specifications

Buffer 6 x DNA Loading Buffer (OXG)
Color Blue, Orange
Concentration 6 x Loading Dye
Form Liquid
Quantity 250 μL
Storage Requirements 4°C for short-term use, or -20°C for long-term storage.
Product Type DNA Molecular Weight Marker

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.