missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCR Master Mix Starter Pack

Product Code. 15545053
Change view
Click to view available options
Quantity:
1 pack
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15545053 1 pack 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15545053 Supplier Invitrogen™ Supplier No. A31021
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The PCR Master Mix Starter Pack is composed of three different PCR master mixes for easy evaluation of their suitability for use in your research.

The PCR Master Mix Starter Pack is composed of three different PCR master mixes for easy evaluation of their suitability for use in your research. These master mixes provide our best DNA polymerases for routine PCR, hot-start PCR, and high-fidelity PCR needs. Thermo Scientific DreamTaq Green PCR Master Mix contains an enhanced Taq DNA polymerase optimized for all standard PCR applications. Invitrogen Platinum II Hot Start PCR Master Mix contains hot-start Taq DNA polymerase for specific, robust, and reliable PCR. Invitrogen SuperFi II PCR Master Mix features superior performance and an error rate 300 times lower than that of Taq. It is an excellent choice for cloning, mutagenesis, and other applications requiring high fidelity.

Master mixes included in this starter pack:

  • 2X DreamTaq Green PCR Master Mix (2X), 200 reactions (Cat. No. K1081)
  • 2X Platinum II Hot Start PCR Master Mix (2X), 50 reactions (14000012)
  • Platinum SuperFi II PCR Master Mix, 100 reactions (12368010)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 2X DreamTaq Green PCR Master Mix, 200 reactions
• 2X Platinum II Hot-Start PCR Master Mix, 50 reactions
• 2X Platinum SuperFi II PCR Master Mix, 100 reactions

Store all components at -20°C.
Polymerase Platinum II Taq Hot Start DNA Polymerase
Kit Type PCR Master Mix Starter Pack
Product Type PCR Master Mix Starter Pack
Quantity 1 pack
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
No. of Reactions 350 Reactions
Reaction Format Kit
Starting Material DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.