missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ PCR Nucleotide Mix

Product Code. 10163103
Change view
Click to view available options
Quantity:
200 μL
1000 μL
Unit Size:
1mL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10163103 200 μL 200µL
10091743 1000 μL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10163103 Supplier Fisher BioReagents™ Supplier No. BP25652
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A premixed 10mM solution of dATP, dCTP, dGTP, and dTTP used for PCR

  • Functionally tested in PCR
  • High purity: >98% triphosphate
  • Supplied as 10mM solution in purified water (pH 7.5)

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
pH 7.5
Content And Storage -20°C
Concentration 10 mM
Purity >98%
Quantity 200 μL
Color Colorless
For Use With (Application) PCR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.