missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ PCR Tube Storage Racks

Product Code. 12587857 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Fluorescent Blue, Green, Red, Violet, Yellow
Fluorescent Yellow
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12587857 Fluorescent Yellow Pack of 25
12577857 Fluorescent Blue, Green, Red, Violet, Yellow Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12587857 Supplier Axygen™ Supplier No. R96PCRFY
  • ONLINE EXCLUSIVE PRICE
    226.00 €
    138.67 € / Pack of 25
    Save 87.33 €
    38% Off
Estimated Shipment: 04-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Constructed of polypropylene. Corning™ Axygen™ PCR Tube Storage Racks hold 96 0.2mL PCR tubes. Supplied with lids.

Polypropylene rack for PCR Tubes

  • Holds 96 0.2mL PCR tubes
  • Available in several colors
  • Supplied with lids
  • Packaged five per pack and five packs per case. Spectrum pack includes five racks of each fluorescent color
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Fluorescent Yellow
Holds Microtubes (0.2 to 0.5 mL)
Material Polycarbonate
Positions 96
No. per Case 25/Cs.
No. per Pack 5/Pk.
Quantity 25/Cs.
Includes Clear Polycarbonate Cover
Product Type Microtube Rack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.