missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ PCR Tubes

Product Code. 16675472
Change view
Click to view available options
Volume (Metric):
0.2 mL
0.5 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility unitSize
16675472 0.2 mL Sterile Pack of 1000
16655472 0.2 mL Non-sterile Pack of 1000
16665472 0.5 mL Non-sterile Pack of 1000
16685472 0.5 mL Sterile Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16675472 Supplier Microspec™ Supplier No. AK3
  • ONLINE EXCLUSIVE PRICE
    55.46 €
    50.45 € / Pack of 1000
    Save 5.01 €
    9% Off
In Stock
Add to basket
Edge
Add to basket

PCR Tubes available in EtO sterile or non-sterile.

  • PCR tubes
  • Natural
  • Pack of 1000

Specifications

Closure Color Clear
Color Clear
Volume (Metric) 0.2 mL
Storage Requirements Room Temperature
Quantity 1000
Closure Material Polypropylene
Shape Conical
Sterility Sterile
Material Polypropylene
Dnase Free No
Closure Type Flip Cap
Autoclavable Not Autoclavable
Closure Included Yes
Disposable Single-use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.