missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems PCSK9, Biotinylated, LDL R Inhibitor Screening ELISA Assay Pair

Product Code. 16427499
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests

Product Code. 16427499

Brand: ACROBiosystems EP10396tests

Please to purchase this item. Need a web account? Register with us today!

This pair is useful for screening for inhibitors of human PCSK9 binding to human LDL R

PCSK9 is a crucial protein in the regulation of plasma cholesterol homeostasis. It binds low density lipoprotein receptor (LDL R) to enhance the degradation of LDL R. Therefore, inhibition of PCSK9 has been considered a promising strategy to prevent the receptor from being degraded and promote removal of LDL cholesterol from circulation.

Specifications

Product Type Inhibitory Kit
Kit Contents This inhibitor screening ELISA pair is designed to facilitate the identification and characterization of new PCSK9 pathway inhibitors. This assay employs a simple colorimetric ELISA platform, which measures the binding between immobilized human LDL R and
Quantity 96 Tests
Shipping Condition RT
Storage Requirements -20°C, aviod light
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.